1-Bromo-2,5-difluoro-4-nitrobenzene ≥98.0% (by GC)

Supplier: TCI America

B3926-1G B3926-5G
TCB3926-1GEA 49.61 CAD
TCB3926-1G TCB3926-5G
1-Bromo-2,5-difluoro-4-nitrobenzene ≥98.0% (by GC)
1-Bromo-2,5-difluoro-4-nitrobenzene

CAS Number: 167415-27-2
MDL Number: MFCD00042184
Molecular Formula: C6H2BrF2NO2
Molecular Weight: 237.99
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 58


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 1-Bromo-2,5-difluoro-4-nitrobenzene

Formula: C₆H₂BrF₂NO₂
Melting Pt: 58 °C
MDL Number: MFCD00042184
CAS Number: 167415-27-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=09799643d39f64169e0fb7317dbfc277b5bb5b85&vid=09799643d39f64169e0fb7317dbfc277b5bb5b85&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR