Bis(1,2,2,6,6-pentamethyl-4-piperidyl)sebacate ≥95.0% (by GC)

Supplier: TCI America

Synonyms: Bis(1,2,2,6,6-pentamethylpiperidin-4-yl) decanedioate, Decanedioic Acid Bis(1,2,2,6,6-pentamethyl-4-piperidyl) Ester, Sebacic Acid Bis(1,2,2,6,6-pentamethyl-4-piperidyl) Ester

B3924-25G B3924-5G
TCB3924-25GEA 301.78 CAD
TCB3924-25G TCB3924-5G
Bis(1,2,2,6,6-pentamethyl-4-piperidyl)sebacate ≥95.0% (by GC)
Bis(1,2,2,6,6-pentamethyl-4-piperidyl)sebacate

CAS Number: 41556-26-7
MDL Number: MFCD00134706
Molecular Formula: C30H56N2O4
Molecular Weight: 508.79
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 222
Specific Gravity (20/20): 1.00


Product Categories: Materials Chemistry, Resin Additives
Other Product Names / Synonyms: Sebacic Acid Bis(1,2,2,6,6-pentamethyl-4-piperidyl) Ester, Decanedioic Acid Bis(1,2,2,6,6-pentamethyl-4-piperidyl) Ester

Formula: C₃₀H₅₆N₂O₄
MW: 508.79 g/mol
Boiling Pt: 222 °C
Density: 1.00 (20 °C)
MDL Number: MFCD00134706
CAS Number: 41556-26-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d567f13e354d4891f977d3b5e7d2b2b321b456f5&vid=d567f13e354d4891f977d3b5e7d2b2b321b456f5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR