3-Bromosalicylaldehyde ≥98.0%
Supplier: TCI America
Synonyms:
3-Bromo-2-hydroxybenzaldehyde
B3922-1G
B3922-5G
TCB3922-1GEA
55.3
CAD
TCB3922-1G
TCB3922-5G
3-Bromosalicylaldehyde ≥98.0%
3-Bromosalicylaldehyde
CAS Number: 1829-34-1
MDL Number: MFCD00016587
Molecular Formula: C7H5BrO2
Molecular Weight: 201.02
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 56
Flash Point (°C): 110
Formula:
C₇H₅BrO₂ Melting Pt: 56 °C Flash Pt: 110 °C |
MDL Number:
MFCD00016587 CAS Number: 1829-34-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...