1H-Benzotriazole-1-carboxaldehyde ≥95.0% (by GC)

Supplier: TCI America

B3920-1G B3920-5G
TCB3920-1GEA 167.48 CAD
TCB3920-1G TCB3920-5G
1H-Benzotriazole-1-carboxaldehyde ≥95.0% (by GC)
1H-Benzotriazole-1-carboxaldehyde

CAS Number: 72773-04-7
MDL Number: MFCD00239420
Molecular Formula: C7H5N3O
Molecular Weight: 147.14
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Melting point (°C): 97
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Formylation
Other Product Names / Synonyms: 1-Formyl-1H-benzotriazole

Formula: C₇H₅N₃O
Melting Pt: 97 °C
MDL Number: MFCD00239420
CAS Number: 72773-04-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=377126ca4ecbc5465f4905b3fa0c7f851544bf00&vid=377126ca4ecbc5465f4905b3fa0c7f851544bf00&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR