O-tert-Butyl-L-serine ≥97.0%
Supplier: TCI America
Synonyms:
H-Ser(tBu)-OH
B3917-1G
B3917-5G
TCB3917-1GEA
71.1
CAD
TCB3917-1G
TCB3917-5G
O-tert-Butyl-L-serine ≥97.0%
O-tert-Butyl-L-serine
CAS Number: 18822-58-7
MDL Number: MFCD00066088
Molecular Formula: C7H15NO3
Molecular Weight: 161.20
Purity/Analysis Method: >97.0% (HPLC,T)
Form: Crystal
Melting point (°C): 226
Specific rotation [a]20/D: -17 deg (C=1, H2O)
Product Categories: Bioscience, Side Chain Protected Amino Acids (N,C-termini Free)
Other Product Names / Synonyms: H-Ser(tBu)-OH
Formula:
C₇H₁₅NO₃ MW: 161.20 g/mol Melting Pt: 226 °C |
MDL Number:
MFCD00066088 CAS Number: 18822-58-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...