3-Bromo-9-phenyl-9H-carbazole ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
3-Bromo-9-phenylcarbazole
B3908-1G
B3908-5G
TCB3908-1GEA
33.18
CAD
TCB3908-1G
TCB3908-5G
3-Bromo-9-phenyl-9H-carbazole ≥98.0% (by GC)
3-Bromo-9-phenyl-9H-carbazole
CAS Number: 1153-85-1
MDL Number: MFCD11977305
Molecular Formula: C18H12BrN
Molecular Weight: 322.21
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 79
Lambda max.: 352 nm (CH3CN)
Formula:
C18H12BrN MW: 322.20 g/mol Melting Pt: 79 °C |
MDL Number:
MFCD11977305 CAS Number: 1153-85-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...