1-(4-Bromophenyl)pyrrolidine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: N-(4-Bromophenyl)pyrrolidine

B3900-1G B3900-5G
TCB3900-1GEA 61.62 CAD
TCB3900-1G TCB3900-5G
1-(4-Bromophenyl)pyrrolidine ≥98.0% (by GC, titration analysis)
1-(4-Bromophenyl)pyrrolidine

CAS Number: 22090-26-2
MDL Number: MFCD04112480
Molecular Formula: C10H12BrN
Molecular Weight: 226.12
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 107

Formula: C₁₀H₁₂BrN
MW: 226.1130 g/mol
Melting Pt: 107 °C
MDL Number: MFCD04112480
CAS Number: 22090-26-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1cd324f46005ebea0df48605bd54c4ef77eb6ff5&vid=1cd324f46005ebea0df48605bd54c4ef77eb6ff5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR