4-((tert-Butoxycarbonyl)amino)benzoic acid ≥98.0%

Supplier: TCI America

Synonyms: Boc-4-Abz-OH, 4-(Boc-amino)benzoic Acid

B3898-25G B3898-5G
TCB3898-25GEA 167.48 CAD
TCB3898-25G TCB3898-5G
4-((tert-Butoxycarbonyl)amino)benzoic acid ≥98.0%
4-((tert-Butoxycarbonyl)amino)benzoic acid

CAS Number: 66493-39-8
MDL Number: MFCD00037428
Molecular Formula: C12H15NO4
Molecular Weight: 237.26
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 196


Other Product Names / Synonyms: 4-(Boc-amino)benzoic Acid

Formula: C₁₂H₁₅NO₄
MW: 237.3 g/mol
Melting Pt: 196 °C
MDL Number: MFCD00037428
CAS Number: 66493-39-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7e86a2fa4ac08190ab4e35409c79203908283961&vid=7e86a2fa4ac08190ab4e35409c79203908283961&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR