2-Bromo-9,9-dimethylfluorene ≥98.0% (by GC)

Supplier: TCI America

B3894-25G B3894-5G
TCB3894-25GEA 176.8 CAD
TCB3894-25G TCB3894-5G
2-Bromo-9,9-dimethylfluorene ≥98.0% (by GC)
2-Bromo-9,9-dimethylfluorene

CAS Number: 28320-31-2
MDL Number: MFCD09261272
Molecular Formula: C15H13Br
Molecular Weight: 273.17
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 190
Melting point (°C): 69

Formula: C15H13Br
MW: 273.17 g/mol
Boiling Pt: 190 °C
Melting Pt: 69 °C
MDL Number: MFCD09261272
CAS Number: 28320-31-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9f852196420c3b0f7c23f1ba7e410376b2a74deb&vid=9f852196420c3b0f7c23f1ba7e410376b2a74deb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR