3-Bromophenyl methyl sulfone ≥98.0% (by GC)

Supplier: TCI America

B3893-25G B3893-5G
TCB3893-25GEA 303.36 CAD
TCB3893-25G TCB3893-5G
3-Bromophenyl methyl sulfone ≥98.0% (by GC)
3-Bromophenyl methyl sulfone

CAS Number: 34896-80-5
MDL Number: MFCD03095326
Molecular Formula: C7H7BrO2S
Molecular Weight: 235.10
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 105

Formula: C₇H₇BrO₂S
Melting Pt: 105 °C
MDL Number: MFCD03095326
CAS Number: 34896-80-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=06fb96a40178e91cc836bfbc097ce251de847ead&vid=d55b82c0006273d538952e786e03ba2c31c4c32c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR