2-Bromo-4,5-dimethoxybenzoic acid ≥98.0%

Supplier: TCI America

Synonyms: 6-Bromoveratric acid

B3891-1G B3891-5G
TCB3891-1GEA 47.4 CAD
TCB3891-1G TCB3891-5G
2-Bromo-4,5-dimethoxybenzoic acid ≥98.0%
2-Bromo-4,5-dimethoxybenzoic acid

CAS Number: 6286-46-0
MDL Number: MFCD00017542
Molecular Formula: C9H9BrO4
Molecular Weight: 261.07
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 186


Other Product Names / Synonyms: 6-Bromoveratric Acid

Formula: C₉H₉BrO₄
MW: 261.07
Melting Pt: 186 °C
MDL Number: MFCD00017542
CAS Number: 6286-46-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a362345ecfe33ec7df6719dde1a463ed40fd2c3c&vid=a362345ecfe33ec7df6719dde1a463ed40fd2c3c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR