(±)-1-Boc-3-aminopyrrolidine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

B3890-1G B3890-5G
TCB3890-1GEA 135.88 CAD
TCB3890-1G TCB3890-5G
(±)-1-Boc-3-aminopyrrolidine ≥98.0% (by GC, titration analysis)
(±)-1-Boc-3-aminopyrrolidine

CAS Number: 186550-13-0
MDL Number: MFCD01861220
Molecular Formula: C9H18N2O2
Molecular Weight: 186.26
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Specific Gravity (20/20): 1.03
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 1-Boc-3-aminopyrrolidine, 1-(tert-Butoxyarbonyl)-3-aminopyrrolidine, 3-Amino-1-Boc-pyrrolidine

Formula: C₉H₁₈N₂O₂
Density: 1.03 (20 °C)
MDL Number: MFCD01861220
CAS Number: 186550-13-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=50ed788098112ee46708bd685bd75d0af0b6bbf5&vid=50ed788098112ee46708bd685bd75d0af0b6bbf5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR