4-Benzyl-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-aspartate ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
Fmoc-Asp(OBzl)-OH, 4-Benzyl N-Fmoc-L-aspartate, N-Fmoc-L-aspartic Acid 4-Benzyl Ester, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-aspartic Acid 4-Benzyl Ester
CAS Number: 86060-84-6
MDL Number: MFCD00065630
Molecular Formula: C26H23NO6
Molecular Weight: 445.47
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 127
Specific rotation [a]20/D: -20 deg (C=1, DMF)
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Peptide Synthesis, Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: 4-Benzyl N-Fmoc-L-aspartate, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-aspartic Acid 4-Benzyl Ester, N-Fmoc-L-aspartic Acid 4-Benzyl Ester, Fmoc-Asp(OBzl)-OH
Formula:
C₂₆H₂₃NO₆ MW: 445.47 g/mol Melting Pt: 127 °C |
MDL Number:
MFCD00065630 CAS Number: 86060-84-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...