4,7-Bis(5-bromo-2-thienyl)-2,1,3-benzothiadiazole ≥95.0%

Supplier: TCI America

Synonyms: 4,7-Bis(5-bromothiophen-2-yl)benzo[c][1,2,5]thiadiazole

B3886-100MG B3886-1G
TCB3886-100MGEA 99.22 CAD
TCB3886-100MG TCB3886-1G
4,7-Bis(5-bromo-2-thienyl)-2,1,3-benzothiadiazole ≥95.0%
4,7-Bis(5-bromo-2-thienyl)-2,1,3-benzothiadiazole

CAS Number: 288071-87-4
MDL Number: MFCD16619295
Molecular Formula: C14H6Br2N2S3
Molecular Weight: 458.20
Purity/Analysis Method: >95.0% (HPLC,N)
Form: Crystal
Melting point (°C): 247

Formula: C₁₄H₆Br₂N₂S₃
MW: 458.20 g/mol
Melting Pt: 247 °C
MDL Number: MFCD16619295
CAS Number: 288071-87-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=61f3eba5500df6cddc52ba4725c697e7e8cd0faa&vid=61f3eba5500df6cddc52ba4725c697e7e8cd0faa&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR