5-Bromoindoline ≥98.0%

Supplier: TCI America

B3885-1G B3885-5G
TCB3885-1GEA 39.5 CAD
TCB3885-1G TCB3885-5G
5-Bromoindoline ≥98.0%
5-Bromoindoline

CAS Number: 22190-33-6
MDL Number: MFCD00027410
Molecular Formula: C8H8BrN
Molecular Weight: 198.06
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 132
Melting point (°C): 41
Storage Temperature: <0°C

Formula: C₈H₈BrN
MW: 198.06 g/mol
Boiling Pt: 132 °C
Melting Pt: 41 °C
MDL Number: MFCD00027410
CAS Number: 22190-33-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1a52b883305d3d98bea6633dbe02210988114bfd&vid=1a52b883305d3d98bea6633dbe02210988114bfd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR