2-Bromo-p-toluic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

B3879-1G B3879-5G
TCB3879-1GEA 26.86 CAD
TCB3879-1G TCB3879-5G
2-Bromo-p-toluic acid ≥98.0% (by GC, titration analysis)
2-Bromo-p-toluic acid

CAS Number: 7697-27-0
MDL Number: MFCD00045797
Molecular Formula: C8H7BrO2
Molecular Weight: 215.05
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 143


Other Product Names / Synonyms: 2-Bromo-p-toluic Acid

Formula: C₈H₇BrO₂
Melting Pt: 143 °C
MDL Number: MFCD00045797
CAS Number: 7697-27-0

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR