1-Bromo-4-hexyloxybenzene ≥98.0% (by GC)
Supplier: TCI America
B3876-25G
B3876-5G
TCB3876-25GEA
246.48
CAD
TCB3876-25G
TCB3876-5G
1-Bromo-4-hexyloxybenzene ≥98.0% (by GC)
1-Bromo-4-hexyloxybenzene
CAS Number: 30752-19-3
MDL Number: MFCD00173753
Molecular Formula: C12H17BrO
Molecular Weight: 257.17
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 154
Specific Gravity (20/20): 1.23
Other Product Names / Synonyms: 1-(4-Bromophenoxy)hexane, 4-Bromophenyl Hexyl Ether
Formula:
C₁₂H₁₇BrO Boiling Pt: 154 °C Density: 1.23 (20 °C) |
MDL Number:
MFCD00173753 CAS Number: 30752-19-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...