2-Bromo-5-fluoroanisole ≥95.0% (by GC)

Supplier: TCI America

Synonyms: 1-Bromo-4-fluoro-2-methoxybenzene

B3875-25G B3875-5G
TCB3875-25GEA 699.94 CAD
TCB3875-25G TCB3875-5G
2-Bromo-5-fluoroanisole ≥95.0% (by GC)
2-Bromo-5-fluoroanisole

CAS Number: 450-88-4
MDL Number: MFCD04973752
Molecular Formula: C7H6BrFO
Molecular Weight: 205.03
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 208
Flash Point (°C): 90
Specific Gravity (20/20): 1.61


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 1-Bromo-4-fluoro-2-methoxybenzene

Formula: C₇H₆BrFO
MW: 205.03
Boiling Pt: 208 °C
Density: 1.61 (20 °C)
Flash Pt: 90 °C
MDL Number: MFCD04973752
CAS Number: 450-88-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=71fd22de4837c9d99ba95e642264b1a600d615cf&vid=71fd22de4837c9d99ba95e642264b1a600d615cf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR