2-(5-Bromo-2-thienyl)-2,3-dihydro-1H-naphtho[1,8-de][1,3,2]diazaborine ≥98.0%

Supplier: TCI America

Synonyms: 2-Bromo-5-(2,3-dihydro-1H-naphtho[1,8-de]-1,3,2-diazaborinyl)thiophene, 2-(5-Bromo-2-thienyl)-2,3-dihydro-1H-naphtho[1,8-de][1,3,2]diazaborinine

B3868-1G B3868-5G
TCB3868-1GEA 148.84 CAD
TCB3868-1G TCB3868-5G
2-(5-Bromo-2-thienyl)-2,3-dihydro-1H-naphtho[1,8-de][1,3,2]diazaborine ≥98.0%
2-(5-Bromo-2-thienyl)-2,3-dihydro-1H-naphtho[1,8-de][1,3,2]diazaborine

CAS Number: 927384-46-1
Molecular Formula: C14H10BBrN2S
Molecular Weight: 329.02
Purity/Analysis Method: >98.0% (GC,N)
Form: Crystal
Melting point (°C): 141
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Boronic Acids & Boronic Acid Derivatives, Materials Chemistry, Conducting Polymer Research Reagents
Other Product Names / Synonyms: 2-Bromo-5-(2,3-dihydro-1H-naphtho[1,8-de]-1,3,2-diazaborinyl)thiophene, 2-(5-Bromo-2-thienyl)-2,3-dihydro-1H-naphtho[1,8-de][1,3,2]diazaborinine

Formula: C₁₄H₁₀BBrN₂S
MW: 329.02 g/mol
Melting Pt: 141 °C
CAS Number: 927384-46-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f3532d60f0790eeb1978c4c56f676c9755ea7295&vid=f3532d60f0790eeb1978c4c56f676c9755ea7295&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR