2-Bromo-3-hexyl-5-iodothiophene ≥97.0% (by GC) stabilized

Supplier: TCI America

B3865-1G B3865-5G
TCB3865-1GEA 165.9 CAD
TCB3865-1G TCB3865-5G
2-Bromo-3-hexyl-5-iodothiophene ≥97.0% (by GC) stabilized
2-Bromo-3-hexyl-5-iodothiophene

(stabilized with Copper chip)
CAS Number: 160096-76-4
MDL Number: MFCD15144651
Molecular Formula: C10H14BrIS
Molecular Weight: 373.09
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 121
Specific Gravity (20/20): 1.71
Storage Temperature: <0°C

Formula: C₁₀H₁₄BrIS
Boiling Pt: 121 °C
Density: 1.71 (20 °C)
MDL Number: MFCD15144651
CAS Number: 160096-76-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b56441cb5e3d5265567104bbbe4233e4152c611c&vid=b56441cb5e3d5265567104bbbe4233e4152c611c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR