2,3-Dihydrobenzo[b][1,4]dioxine-5-carboxylic acid ≥98.0%

Supplier: TCI America

Synonyms: 2,3-dihydro-1,4-benzodioxine-5-carboxylic acid, 2,3-dihydrobenzo[b][1,4]dioxine-5-carboxylic acid, 1,4-Benzodioxan-5-carboxylic acid

B3864-25G B3864-5G
TCB3864-25GEA 308.1 CAD
TCB3864-25G TCB3864-5G
2,3-Dihydrobenzo[b][1,4]dioxine-5-carboxylic acid ≥98.0%
2,3-Dihydrobenzo[b][1,4]dioxine-5-carboxylic acid

CAS Number: 4442-53-9
MDL Number: MFCD00239415
Molecular Formula: C9H8O4
Molecular Weight: 180.16
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 192


Other Product Names / Synonyms: 2,3-Ethylenedioxybenzoic Acid

Formula: C₉H₈O₄
Melting Pt: 192 °C
MDL Number: MFCD00239415
CAS Number: 4442-53-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=318112db7ebb15e728f958a3e300fdf1d47ab494&vid=318112db7ebb15e728f958a3e300fdf1d47ab494&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR