2-Bromo-3-methoxypyridine ≥98.0% (by GC)
Supplier: TCI America
B3861-25G
B3861-5G
TCB3861-25GEA
518.24
CAD
TCB3861-25G
TCB3861-5G
2-Bromo-3-methoxypyridine ≥98.0% (by GC)
2-Bromo-3-methoxypyridine
CAS Number: 24100-18-3
MDL Number: MFCD01570896
Molecular Formula: C6H6BrNO
Molecular Weight: 188.02
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 47
Flash Point (°C): 110
Formula:
C₆H₆BrNO Melting Pt: 47 °C Flash Pt: 110 °C |
MDL Number:
MFCD01570896 CAS Number: 24100-18-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...