2-Bromo-3-chloropyridine ≥98.0% (by GC)
Supplier: TCI America
B3860-25G
B3860-5G
TCB3860-25GEA
131.14
CAD
TCB3860-25G
TCB3860-5G
2-Bromo-3-chloropyridine ≥98.0% (by GC)
2-Bromo-3-chloropyridine
CAS Number: 96424-68-9
MDL Number: MFCD04039344
Molecular Formula: C5H3BrClN
Molecular Weight: 192.44
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 60
Formula:
C₅H₃BrClN Melting Pt: 60 °C |
MDL Number:
MFCD04039344 CAS Number: 96424-68-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...