3-Benzyloxybenzyl alcohol ≥97.0%
Supplier: TCI America
Synonyms:
(3-(Benzyloxy)phenyl)methanol
B3857-25G
B3857-5G
TCB3857-25GEA
461.36
CAD
TCB3857-25G
TCB3857-5G
3-Benzyloxybenzyl alcohol ≥97.0%
3-Benzyloxybenzyl alcohol
CAS Number: 1700-30-7
MDL Number: MFCD00004642
Molecular Formula: C14H14O2
Molecular Weight: 214.26
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 160
Melting point (°C): 49
Formula:
C₁₄H₁₄O₂ Boiling Pt: 160 °C Melting Pt: 49 °C |
MDL Number:
MFCD00004642 CAS Number: 1700-30-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...