4-(Boc-amino)cyclohexanone ≥98.0%

Supplier: TCI America

Synonyms: 4-Aminocyclohexan-1-one, N-BOC protected, tert-Butyl (4-oxocyclohexyl)carbamate

B3856-1G B3856-5G
TCB3856-1GEA 53.72 CAD
TCB3856-1G TCB3856-5G
4-(Boc-amino)cyclohexanone ≥98.0%
4-(Boc-amino)cyclohexanone

CAS Number: 179321-49-4
MDL Number: MFCD00798168
Molecular Formula: C11H19NO3
Molecular Weight: 213.28
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 116


Other Product Names / Synonyms: 4-(Boc-amino)cyclohexanone

Formula: C₁₁H₁₉NO₃
MW: 213.2700 g/mol
Melting Pt: 116 °C
MDL Number: MFCD00798168
CAS Number: 179321-49-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=08ca80c70f595b2306eb73d56e9911a1f58f12da&vid=08ca80c70f595b2306eb73d56e9911a1f58f12da&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR