1,3-Benzenedimethanethiol ≥98.0%

Supplier: TCI America

Synonyms: 1,3-Bis(mercaptomethyl)benzene, alpha,alpha'-Dimercapto-m-xylene

B3854-1G B3854-5G
TCB3854-1GEA 123.24 CAD
TCB3854-1G TCB3854-5G
1,3-Benzenedimethanethiol ≥98.0%
1,3-Benzenedimethanethiol

CAS Number: 41563-69-3
MDL Number: MFCD00012311
Molecular Formula: C8H10S2
Molecular Weight: 170.29
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 170
Flash Point (°C): 113
Specific Gravity (20/20): 1.16


Product Categories: Synthetic Organic Chemistry, Thiols
Other Product Names / Synonyms: 1,3-Bis(mercaptomethyl)benzene, alpha,alpha'-Dimercapto-m-xylene

Formula: C₈H₁₀S₂
Boiling Pt: 170 °C
Density: 1.16 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00012311
CAS Number: 41563-69-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a22b0a41d71203f264a7407b1dc9074663e29ddc&vid=a22b0a41d71203f264a7407b1dc9074663e29ddc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR