3-Phenylbenzoic acid ≥98.0%

Supplier: TCI America

Synonyms: Biphenyl-3-carboxylic acid, 3-Biphenylcarboxylic acid, 1,1'-Biphenyl-3-carboxylic acid, [1,1'-Biphenyl]-3-carboxylic acid

B3850-1G B3850-5G
TCB3850-1GEA 38.01 CAD
TCB3850-1G TCB3850-5G
3-Phenylbenzoic acid ≥98.0%
3-Phenylbenzoic acid

CAS Number: 716-76-7
MDL Number: MFCD00045846
Molecular Formula: C13H10O2
Molecular Weight: 198.22
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 166


Other Product Names / Synonyms: 3-Phenylbenzoic Acid

Formula: C₁₃H₁₀O₂
MW: 198.22 g/mol
Melting Pt: 166 °C
MDL Number: MFCD00045846
CAS Number: 716-76-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5d6e0a8573d1b319c389a8bea31f863f2cb064f0&vid=5d6e0a8573d1b319c389a8bea31f863f2cb064f0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR