3-Bromo-2,4,6-trimethylaniline ≥98.0% (by GC, titration analysis)

Supplier: TCI America

B3849-25G B3849-5G
TCB3849-25GEA 1715.88 CAD
TCB3849-25G TCB3849-5G
3-Bromo-2,4,6-trimethylaniline ≥98.0% (by GC, titration analysis)
3-Bromo-2,4,6-trimethylaniline

CAS Number: 82842-52-2
MDL Number: MFCD00015476
Molecular Formula: C9H12BrN
Molecular Weight: 214.11
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 173
Melting point (°C): 38
Flash Point (°C): 113
Storage Temperature: 0-10°C

Formula: C₉H₁₂BrN
Boiling Pt: 173 °C
Melting Pt: 38 °C
Flash Pt: 113 °C
MDL Number: MFCD00015476
CAS Number: 82842-52-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d4ea2f5bbe5df4647b24d1a4b64f6e07103de1b9&vid=d4ea2f5bbe5df4647b24d1a4b64f6e07103de1b9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR