1-Bromo-3,5-dimethoxybenzene ≥98.0% (by GC)

Supplier: TCI America

B3848-25G B3848-5G
TCB3848-25GEA 107.44 CAD
TCB3848-25G TCB3848-5G
1-Bromo-3,5-dimethoxybenzene ≥98.0% (by GC)
1-Bromo-3,5-dimethoxybenzene

CAS Number: 20469-65-2
MDL Number: MFCD06200685
Molecular Formula: C8H9BrO2
Molecular Weight: 217.06
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 67

Formula: C₈H₉BrO₂
MW: 217.06 g/mol
Melting Pt: 67 °C
MDL Number: MFCD06200685
CAS Number: 20469-65-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8b2a50fcb38a45604ca8d3b494642325f9a47815&vid=8b2a50fcb38a45604ca8d3b494642325f9a47815&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR