2-Bromo-5-methoxybenzoic acid ≥98.0%

Supplier: TCI America

B3846-25G B3846-5G
TCB3846-25GEA 132.72 CAD
TCB3846-25G TCB3846-5G
2-Bromo-5-methoxybenzoic acid ≥98.0%
2-Bromo-5-methoxybenzoic acid

CAS Number: 22921-68-2
MDL Number: MFCD00020214
Molecular Formula: C8H7BrO3
Molecular Weight: 231.05
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 161


Other Product Names / Synonyms: 6-Bromo-m-anisic Acid

Formula: C₈H₇BrO₃
Melting Pt: 161 °C
MDL Number: MFCD00020214
CAS Number: 22921-68-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=75b776264653aa2a695a6fee52c019cf8105bdf2&vid=f41fe291d9145e6a6f51353ae1ebeca0bb3c04aa&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR