2-Bromo-4,5-difluorophenol ≥98.0% (by GC)

Supplier: TCI America

B3845-5G B3845-1G
TCB3845-5GEA 105.86 CAD
TCB3845-5G TCB3845-1G
2-Bromo-4,5-difluorophenol ≥98.0% (by GC)
2-Bromo-4,5-difluorophenol

CAS Number: 166281-37-4
MDL Number: MFCD00070751
Molecular Formula: C6H3BrF2O
Molecular Weight: 208.99
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Flash Point (°C): 81
Specific Gravity (20/20): 1.84

Formula: C₆H₃BrF₂O
Density: 1.84 (20 °C)
Flash Pt: 81 °C
MDL Number: MFCD00070751
CAS Number: 166281-37-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9887993998aba2650dbf0e22f75908ef6a212507&vid=9887993998aba2650dbf0e22f75908ef6a212507&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR