N-(tert-Butoxycarbonyl)-trans-4-hydroxy-L-prolinemethyl ester ≥95.0%

Supplier: TCI America

Synonyms: N-Boc-trans-4-hydroxy-L-proline Methyl Ester, (2S,4R)-4-Hydroxypyrrolidine-1,2-dicarboxylic acid 1-tert-Butyl 2-Methyl Ester, 1-tert-Butyl 2-Methyl (2S,4R)-4-Hydroxypyrrolidine-1,2-dicarboxylate, Boc-Hyp-OMe, 1-(tert-Butyl) 2-methyl (2S,4R)-4-hydroxypyrrolidine-1,2-dicarboxylate

B3843-1G B3843-5G
TCB3843-1GEA 48.98 CAD
TCB3843-1G TCB3843-5G
N-(tert-Butoxycarbonyl)-trans-4-hydroxy-L-prolinemethyl ester ≥95.0%
N-(tert-Butoxycarbonyl)-trans-4-hydroxy-L-prolinemethyl ester

CAS Number: 74844-91-0
MDL Number: MFCD00076981
Molecular Formula: C11H19NO5
Molecular Weight: 245.28
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Boiling point (°C): 132
Melting point (°C): 96
Specific rotation [a]20/D: -79 deg (C=1, MeOH)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks, Bioscience, Amino Acids (N-Protected), Amino Acids (C-Protected)
Other Product Names / Synonyms: N-Boc-trans-4-hydroxy-L-proline Methyl Ester, Boc-Hyp-OMe, 1-tert-Butyl 2-Methyl (2S,4R)-4-Hydroxypyrrolidine-1,2-dicarboxylate, (2S,4R)-4-Hydroxypyrrolidine-1,2-dicarboxylic Acid 1-tert-Butyl 2-Methyl Ester

Formula: C₁₁H₁₉NO₅
MW: 245.28 g/mol
Boiling Pt: 132 °C
Melting Pt: 96 °C
MDL Number: MFCD00076981
CAS Number: 74844-91-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=80c41e48e2707a311ddbedc14bf7470833d881a9&vid=80c41e48e2707a311ddbedc14bf7470833d881a9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR