(2-Bromophenyl)acetone ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 1-(2-Bromophenyl)propan-2-one

B3842-1G B3842-5G
TCB3842-1GEA 45.82 CAD
TCB3842-1G TCB3842-5G
(2-Bromophenyl)acetone ≥98.0% (by GC)
(2-Bromophenyl)acetone

CAS Number: 21906-31-0
Molecular Formula: C9H9BrO
Molecular Weight: 213.07
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 92
Specific Gravity (20/20): 1.43


Other Product Names / Synonyms: 1-(2-Bromophenyl)-2-propanone

Formula: C₉H₉BrO
MW: 213.07 g/mol
Boiling Pt: 92 °C
Density: 1.43 (20 °C)
MDL Number: MFCD03410431
CAS Number: 21906-31-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e056d6ee370a870824419f5b65265426bf739e68&vid=e056d6ee370a870824419f5b65265426bf739e68&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR