2-Bromo-5-nitrobenzoic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
B3841-25G
B3841-5G
TCB3841-25GEA
309.68
CAD
TCB3841-25G
TCB3841-5G
2-Bromo-5-nitrobenzoic acid ≥98.0% (by GC, titration analysis)
2-Bromo-5-nitrobenzoic acid
CAS Number: 943-14-6
MDL Number: MFCD00134558
Molecular Formula: C7H4BrNO4
Molecular Weight: 246.02
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 182
Formula:
C₇H₄BrNO₄ Melting Pt: 182 °C |
MDL Number:
MFCD00134558 CAS Number: 943-14-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...