(S)-2-((tert-Butoxycarbonyl)amino)-2-cyclohexylacetic acid ≥98.0%

Supplier: TCI America

Synonyms: N-Boc-2-cyclohexyl-L-glycine, Boc-cyclohexyl-Gly-OH, BOC-L-α-cyclohexylglycine, N-Boc-L-2-cyclohexylglycine, Boc-Chg-OH

B3839-1G B3839-5G
TCB3839-1GEA 33.18 CAD
TCB3839-1G TCB3839-5G
(S)-2-((tert-Butoxycarbonyl)amino)-2-cyclohexylacetic acid ≥98.0%
(S)-2-((tert-Butoxycarbonyl)amino)-2-cyclohexylacetic acid

CAS Number: 109183-71-3
MDL Number: MFCD00133638
Molecular Formula: C13H23NO4
Molecular Weight: 257.33
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 83
Specific rotation [a]20/D: 11 deg (C=1, MeOH)


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: Boc-Chg-OH, N-Boc-L-2-cyclohexylglycine

Formula: C13H23NO4
MW: 257.3 g/mol
Melting Pt: 83 °C
MDL Number: MFCD00133638
CAS Number: 109183-71-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=24fa1f1e8a6bb69cf9bb01741578510566ea43e7&vid=24fa1f1e8a6bb69cf9bb01741578510566ea43e7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR