5-Bromo-2-(methylthio)pyrimidine-4-carboxylic acid ≥97.0% (by titrimetric analysis)

Supplier: TCI America

B3829-1G B3829-5G
TCB3829-1GEA 42.66 CAD
TCB3829-1G TCB3829-5G
5-Bromo-2-(methylthio)pyrimidine-4-carboxylic acid ≥97.0% (by titrimetric analysis)
5-Bromo-2-(methylthio)pyrimidine-4-carboxylic acid

CAS Number: 50593-92-5
MDL Number: MFCD00238866
Molecular Formula: C6H5BrN2O2S
Molecular Weight: 249.08
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 177

Formula: C₆H₅BrN₂O₂S
Melting Pt: 177 °C
MDL Number: MFCD00238866
CAS Number: 50593-92-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=64a5e9d7c0ce5692967ad75213e7a2317d52556e&vid=64a5e9d7c0ce5692967ad75213e7a2317d52556e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR