Bis(4-hydroxyphenyl) disulfide ≥98.0%

Supplier: TCI America

Synonyms: 4,4'-Dihydroxydiphenyl disulfide, 4,4'-Dithiodiphenol, 4,4'-Dihydroxydiphenyl disulphide, Bis(4-hydroxyphenyl) Disulfide, Bis(4-hydroxyphenyl) Disulphide

B3827-5G B3827-1G
TCB3827-5GEA 147.1 CAD
TCB3827-5G TCB3827-1G
Bis(4-hydroxyphenyl) disulfide ≥98.0%
Bis(4-hydroxyphenyl) disulphide

CAS Number: 15015-57-3
MDL Number: MFCD00020164
Molecular Formula: C12H10O2S2
Molecular Weight: 250.33
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 152


Other Product Names / Synonyms: 4,4'-Dihydroxydiphenyl Disulfide, 4,4'-Dithiodiphenol

Formula: C₁₂H₁₀O₂S₂
MW: 250.34
Melting Pt: 152 °C
MDL Number: MFCD00020164
CAS Number: 15015-57-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cfe195b1310aa87635f0dc8b81614db3fb6885ca&vid=cfe195b1310aa87635f0dc8b81614db3fb6885ca&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR