6-Bromo-2-naphthoic acid ≥98.0%
Supplier: TCI America
B3823-25G
B3823-5G
TCB3823-25GEA
319.16
CAD
TCB3823-25G
TCB3823-5G
6-Bromo-2-naphthoic acid ≥98.0%
6-Bromo-2-naphthoic acid
CAS Number: 5773-80-8
MDL Number: MFCD01075720
Molecular Formula: C11H7BrO2
Molecular Weight: 251.08
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 297
Other Product Names / Synonyms: 6-Bromo-2-naphthalenecarboxylic Acid
Formula:
C₁₁H₇BrO₂ Melting Pt: 297 °C |
MDL Number:
MFCD01075720 CAS Number: 5773-80-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...