N,N,N',N'-Bis(tetramethylene)-O-(N-succinimidyl)uronium hexafluorophosphate ≥98.0%

Supplier: TCI America

Synonyms: 1-(((2,5-Dioxopyrrolidin-1-yl)oxy)(pyrrolidin-1-yl)methylene)pyrrolidin-1-ium hexafluorophosphate(V), HSPyU, Dipyrrolidino(N-succinimidyloxy)carbenium Hexafluorophosphate, Dipyrrolidino(N-succinimidyloxy)carbenium hexafluorophosphate

B3817-1G B3817-5G
TCB3817-1GEA 82 CAD
TCB3817-1G TCB3817-5G
N,N,N',N'-Bis(tetramethylene)-O-(N-succinimidyl)uronium hexafluorophosphate ≥98.0%
N,N,N',N'-Bis(tetramethylene)-O-(N-succinimidyl)uronium hexafluorophosphate

CAS Number: 207683-26-9
MDL Number: MFCD00192457
Molecular Formula: C13H20F6N3O3P
Molecular Weight: 411.29
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 143
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Condensation & Active Esterification
Other Product Names / Synonyms: Dipyrrolidino(N-succinimidyloxy)carbenium Hexafluorophosphate, HSPyU

Formula: C₁₃H₂₀F₆N₃O₃P
Melting Pt: 143 °C
MDL Number: MFCD00192457
CAS Number: 207683-26-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dd719586c3ead419025e9b8aff4e1b8141e106ce&vid=dd719586c3ead419025e9b8aff4e1b8141e106ce&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR