(Benzotriazol-1-yloxy)dipiperidinocarbenium hexafluorophosphate ≥98.0% (by HPLC)

Supplier: TCI America

B3816-1G B3816-5G
TCB3816-1GEA 79.32 CAD
TCB3816-1G TCB3816-5G
(Benzotriazol-1-yloxy)dipiperidinocarbenium hexafluorophosphate ≥98.0% (by HPLC)
(Benzotriazol-1-yloxy)dipiperidinocarbenium hexafluorophosphate

CAS Number: 206752-41-2
MDL Number: MFCD00191769
Molecular Formula: C17H24F6N5OP
Molecular Weight: 459.38
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 199
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Condensation & Active Esterification, Peptide Synthesis, Peptide Synthesis
Other Product Names / Synonyms: (Benzotriazol-1-yloxy)dipiperidinocarbenium Hexafluorophosphate, HBPipU

Formula: C₁₇H₂₄F₆N₅OP
Melting Pt: 199 °C
MDL Number: MFCD00191769
CAS Number: 190849-64-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=10ef29786695f778f3434186c34ebeae095054c0&vid=10ef29786695f778f3434186c34ebeae095054c0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR