4-Bromo-2-nitrobenzoic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

B3810-25G B3810-5G
TCB3810-25GEA 282.82 CAD
TCB3810-25G TCB3810-5G
4-Bromo-2-nitrobenzoic acid ≥98.0% (by titrimetric analysis)
4-Bromo-2-nitrobenzoic acid

CAS Number: 99277-71-1
MDL Number: MFCD01013599
Molecular Formula: C7H4BrNO4
Molecular Weight: 246.02
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 165

Formula: C₇H₄BrNO₄
Melting Pt: 165 °C
MDL Number: MFCD01013599
CAS Number: 99277-71-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=294c7b54343cc6fc3a08810531dcca51f0d1c906&vid=294c7b54343cc6fc3a08810531dcca51f0d1c906&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR