1-Bromo-3-fluoro-4-(trifluoromethoxy)benzene ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 4-Bromo-2-fluoro-1-(trifluoromethoxy)benzene

B3808-1G B3808-5G
TCB3808-1GEA 26.86 CAD
TCB3808-1G TCB3808-5G
1-Bromo-3-fluoro-4-(trifluoromethoxy)benzene ≥98.0% (by GC)
1-Bromo-3-fluoro-4-(trifluoromethoxy)benzene

CAS Number: 105529-58-6
MDL Number: MFCD01861132
Molecular Formula: C7H3BrF4O
Molecular Weight: 259.00
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.71


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Fluorinated Building Blocks
Other Product Names / Synonyms: 4-Bromo-2-fluoro-1-(trifluoromethoxy)benzene

Formula: C₇H₃BrF₄O
MW: 259.00 g/mol
Density: 1.71 (20 °C)
MDL Number: MFCD01861132
CAS Number: 105529-58-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=02a650c2abb5bbe9289035edef7dfab628e3ba20&vid=02a650c2abb5bbe9289035edef7dfab628e3ba20&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR