BMB (1,4-Bis(maleimido)butane) ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
B3805-100MG
B3805-1G
TCB3805-100MGEA
135.56
CAD
TCB3805-100MG
TCB3805-1G
BMB (1,4-Bis(maleimido)butane) ≥98.0% (by HPLC, total nitrogen)
BMB (1,4-Bis(maleimido)butane)
CAS Number: 28537-70-4
MDL Number: MFCD01318342
Molecular Formula: C12H12N2O4
Molecular Weight: 248.24
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 206
Storage Temperature: <0°C
Product Categories: Materials Chemistry, Monomers, Bioscience, Crosslinkers
Other Product Names / Synonyms: N,N'-Tetramethylenebismaleimide, N,N'-Tetramethylenedimaleimide, BMB
Formula:
C₁₂H₁₂N₂O₄ Melting Pt: 206 °C |
MDL Number:
MFCD01318342 CAS Number: 28537-70-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...