1-Boc-3-oxopiperazine ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
tert-butyl 3-oxopiperazine-1-carboxylate, tert-butyl 3-oxo-1-piperazinecarboxylate, 1-Boc-3-Oxopiperazine, 4-N-BOC-2-oxo-piperazine, 4-Boc-2-Oxopiperazine
CAS Number: 76003-29-7
MDL Number: MFCD02181069
Molecular Formula: C9H16N2O3
Molecular Weight: 200.24
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 159
Other Product Names / Synonyms: 1-Boc-3-oxopiperazine, 3-Oxopiperazine-1-carboxylic Acid tert-Butyl Ester, 1-(tert-Butoxycarbonyl)-3-oxopiperazine
Formula:
C₉H₁₆N₂O₃ MW: 200.238 g/mol Melting Pt: 159 °C |
MDL Number:
MFCD02181069 CAS Number: 76003-29-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...