Butein ≥98.0%
Supplier: TCI America
Synonyms:
2',3,4,4'-Tetrahydroxychalcone, (E)-1-(2,4-Dihydroxyphenyl)-3-(3,4-dihydroxyphenyl)-2-propen-1-one
CAS Number: 487-52-5
MDL Number: MFCD00017300
Molecular Formula: C15H12O5
Molecular Weight: 272.26
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 215
Storage Temperature: <0°C
Product Categories: Bioscience, Inhibitors (for Pharmacological Research), Antioxidants, Aromatic Polyketides, Polyphenols (Phytochemicals), Histone Deacetylase Activators
Other Product Names / Synonyms: (E)-1-(2,4-Dihydroxyphenyl)-3-(3,4-dihydroxyphenyl)-2-propen-1-one, 2',3,4,4'-Tetrahydroxychalcone
Formula:
C₁₅H₁₂O₅ Melting Pt: 215 °C |
MDL Number:
MFCD00017300 CAS Number: 487-52-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...