4-Bromo-1-indanone ≥97.0% (by GC)

Supplier: TCI America

B3801-1G B3801-5G
TCB3801-1GEA 33.18 CAD
TCB3801-1G TCB3801-5G
4-Bromo-1-indanone ≥97.0% (by GC)
4-Bromo-1-indanone

CAS Number: 15115-60-3
MDL Number: MFCD01719772
Molecular Formula: C9H7BrO
Molecular Weight: 211.06
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 125
Melting point (°C): 94

Formula: C₉H₇BrO
Boiling Pt: 125 °C
Melting Pt: 94 °C
MDL Number: MFCD01719772
CAS Number: 15115-60-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=421d6a8d2c0cb66f65a2796704f02efa5942d431&vid=421d6a8d2c0cb66f65a2796704f02efa5942d431&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR