2-Bromo-4-(trifluoromethoxy)aniline ≥98.0% (by GC)

Supplier: TCI America

B3799-25G B3799-5G
TCB3799-25GEA 251.22 CAD
TCB3799-25G TCB3799-5G
2-Bromo-4-(trifluoromethoxy)aniline ≥98.0% (by GC)
2-Bromo-4-(trifluoromethoxy)aniline

CAS Number: 175278-17-8
MDL Number: MFCD00203478
Molecular Formula: C7H5BrF3NO
Molecular Weight: 256.02
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 209
Flash Point (°C): 113
Specific Gravity (20/20): 1.70

Formula: C₇H₅BrF₃NO
Boiling Pt: 209 °C
Density: 1.70 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00203478
CAS Number: 175278-17-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=12563bb820920f02aac1fed8716c9fde9cb9d757&vid=12563bb820920f02aac1fed8716c9fde9cb9d757&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR