1-Benzhydrylazetidine-3-carboxylic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

B3794-25G B3794-5G
TCB3794-25GEA 1775.92 CAD
TCB3794-25G TCB3794-5G
1-Benzhydrylazetidine-3-carboxylic acid ≥98.0% (by HPLC, titration analysis)
1-Benzhydrylazetidine-3-carboxylic acid

CAS Number: 36476-87-6
MDL Number: MFCD01566538
Molecular Formula: C17H17NO2
Molecular Weight: 267.33
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 198


Other Product Names / Synonyms: 1-(Diphenylmethyl)azetidine-3-carboxylic Acid

Formula: C₁₇H₁₇NO₂
Melting Pt: 198 °C
MDL Number: MFCD01566538
CAS Number: 36476-87-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cfa06187cc53a2f9f0e49347a67588e0f998854c&vid=cfa06187cc53a2f9f0e49347a67588e0f998854c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR