Benzo[1,2-b:4,5-b']dithiophene-4,8-dione ≥98.0% (by GC)

Supplier: TCI America

B3793-1G B3793-5G
TCB3793-1GEA 200.66 CAD
TCB3793-1G TCB3793-5G
Benzo[1,2-b:4,5-b']dithiophene-4,8-dione ≥98.0% (by GC)
Benzo[1,2-b:4,5-b']dithiophene-4,8-dione

CAS Number: 32281-36-0
Molecular Formula: C10H4O2S2
Molecular Weight: 220.26
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 260

Formula: C₁₀H₄O₂S₂
Melting Pt: 260 °C
MDL Number: MFCD01927240
CAS Number: 32281-36-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=31321b3a82943c26b9a30560bf53da4b08d2a206&vid=31321b3a82943c26b9a30560bf53da4b08d2a206&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR