4-Bromo-3-chlorobenzoic acid ≥98.0%

Supplier: TCI America

B3791-25G B3791-5G
TCB3791-25GEA 303.36 CAD
TCB3791-25G TCB3791-5G
4-Bromo-3-chlorobenzoic acid ≥98.0%
4-Bromo-3-chlorobenzoic acid

CAS Number: 25118-59-6
MDL Number: MFCD08276864
Molecular Formula: C7H4BrClO2
Molecular Weight: 235.46
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 222

Formula: C₇H₄BrClO₂
Melting Pt: 222 °C
MDL Number: MFCD08276864
CAS Number: 25118-59-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=86258ec4db1c99b9bf26f5be64611eb1ae42b133&vid=86258ec4db1c99b9bf26f5be64611eb1ae42b133&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR